Anti-DSG1 Mouse Monoclonal Antibody [clone: CC49]
ABNOMAB12146
: Abnova
- Antibody type:Primary
- Antigen name:Desmoglein 1
- Antigen symbol:DSG1
- Clonality:Monoclonal
- Clone:CC49
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1828
- Antigen synonyms:EPKHE|SPPK1|DG1|PPKS1|EPKHIA|CDHF4|DSG
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide).
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Purified human DSG1 protein.
- Purification:Protein A/G purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against human DSG1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: DSG1
Clonality: Monoclonal
Clone: CC49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: