Order Entry
Denmark
ContactUsLinkComponent
Anti-CCP110 Rabbit Polyclonal Antibody
Anti-CCP110 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25493
Supplier:  Abnova
Anti-CCP110 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25493
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Centriolar coiled coil Protein 110kDa
  • Antigen symbol:
    CCP110
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9738
  • Antigen synonyms:
    Cep110|Centrosomal protein of 110 kDa|KIAA0419|CP110|CEP110|Centriolar coiled-coil protein of 110 kDa
  • Storage buffer:
    PBS (0,02% sodium azide)
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to C-terminus of human CP110.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of CP110.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: CCP110
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat