Anti-JUN Rabbit Polyclonal Antibody
ABNOPAB25222
: Abnova
- Antibody type:Primary
- Antigen name:c-Jun
- Antigen symbol:JUN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3725
- Antigen synonyms:Transcription factor AP-1|V-jun avian sarcoma virus 17 oncogene homolog|JUN|AP1|p39|Proto-oncogene c-jun|Activator protein 1
- Modification:phospho-Ser63
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Sequence:L-T-Sp-P-D
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S63 of human JUN.
- Purification:purified by Affinity Chromatography
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUN.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000
Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat