Anti-LPL Mouse Monoclonal Antibody [clone: 2C5]
Catalog # ABNOMAB10791
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lipoprotein lipase
- Antigen symbol:LPL
- Clonality:Monoclonal
- Clone:2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:4023
- Antigen synonyms:LIPD|LPL protein|LPL|MGC137861|HDLCQ11|MGC93586.|Lipase|lipoprotein lipase|Lipoprotein lipase 1
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human LPL.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant LPL.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: LPL
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human