Anti-FUT4 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25143
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fucosyltransferase 4
- Antigen symbol:FUT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2526
- Antigen synonyms:CD15|FUC-TIV|ELFT|SSEA-1|LeX|FCT3A|FUTIV
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to FUT4.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of FUT4.
Recommended Dilutions: Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: FUT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human