Ordreindtastning
Denmark
ContactUsLinkComponent
 
Anti-CDKN1C Mouse Monoclonal Antibody [clone: 3E3]
Catalog # ABNOMAB10550
Supplier:  Abnova
Anti-CDKN1C Mouse Monoclonal Antibody [clone: 3E3]
Catalog # ABNOMAB10550
Supplier:  Abnova

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase inhibitor 1C
  • Antigen symbol:
    CDKN1C
  • Clonality:
    Monoclonal
  • Clone:
    3E3
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Krydsadsorption:
    No
  • Udformning:
    Liquid
  • Form:
    Ascites
  • Gen ID:
    1028
  • Antigen synonymer:
    BWCR|p57|WBS|p57Kip2|BWS|KIP2
  • Storage buffer:
    0,03% sodium azide
  • Opbevaringstemperatur:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human CDKN1C.
  • Størrelse:
    100 μl
  • Pakn:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant CDKN1C.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: CDKN1C
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human