Anti-CDH1 Mouse Monoclonal Antibody [clone: 7H12]
Catalog # ABNOMAB10546
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cadherin 1
- Antigen symbol:CDH1
- Clonality:Monoclonal
- Clone:7H12
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:999
- Antigen synonyms:CD324|epithelial Cadherin|Cadherin-E|ECAD|Arc-1|CAM 120/80|cell-CAM 120/80|E-Cadherin|uvomorulin|Cadherin 1|Cadherin-1|E-Cadherin epithelial|calcium-dependent adhesion protein|epithelial|LCAM|CDHE|UVO
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CDH1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CDH1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 7H12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse