Anti-KLRD1 Mouse Monoclonal Antibody [clone: 1H1]
Catalog # ABNOMAB10543
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Killer Cell Lectin-like Receptor Subfamily D Member 1
- Antigen symbol:KLRD1
- Clonality:Monoclonal
- Clone:1H1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:3824
- Antigen synonyms:CD94
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human KLRD1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant KLRD1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: KLRD1
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human