Anti-CDH2 Mouse Monoclonal Antibody [clone: 4B11]
Catalog # ABNOH00001000-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cadherin 2
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:4B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1000
- Antigen synonyms:CDHN|Neural-Cadherin|CDw325|N-Cadherin|CDH2|NCAD
- Amino acid number:807 to 906
- Storage buffer:1x PBS, pH 7,4
- Sequence:RRMDERPIHAEPQYPVRSAAPHPGDIGDFINEGLKAADNDPTAPPYDSLLVFDYEGSGSTAGSLSSLNSSSSGGEQDYDYLNDWGPRFKKLADMYGGGDD
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CDH2 (NP_001783, 807 a.a. ~ 906 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant CDH2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 4B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human