- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 1
- Antigen symbol:MAPK1
- Clonality:Monoclonal
- Clone:G15-B
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:5594|5595
- Antigen synonyms:P42MAPK|PRKM2|ERK2|PRKM1|MAPK2|ERK|ERT1|p38|p41mapk|ERK-2|p40|p42-MAPK|p41
- Modification:phospho-Thr202/204
- Storage buffer:20 mM Tris-HCl, pH 8.0 (10 mg/mL BSA, 0,05% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding T202/T204 of human MAPK1/MAPK3.
- Size:50 μl
- Pk:50 µl
Rabbit monoclonal antibody raised against synthetic phosphopeptide of MAPK1/MAPK3.
Recommended Dilutions: Western Blot: 1:5000; ELISA: 1:20000-1:100000
Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: G15-B
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat