Anti-Macrophage/Monocyte Mouse Monoclonal Antibody [clone: RbM2]
Catalog # ABNOMAB1735
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Macrophage/Monocyte
- Clonality:Monoclonal
- Clone:RbM2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,1% proclin, 2.0% Block Ace)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native from Thioglycate-elicited rabbit peritoneal macrophages.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against macrophage/monocyte.
Recommended Dilutions: Immunohistochemistry: 10 ?g/ml
Type: Primary
Antigen: Macrophage/Monocyte
Clonality: Monoclonal
Clone: RbM2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit