Anti-HSPBP1 Rabbit Polyclonal Antibody
ABNOPAB0882
: Abnova
- Antibody type:Primary
- Antigen name:Heat Shock Protein-Binding Protein 1
- Antigen symbol:HSPBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:23640
- Antigen synonyms:Hsp70-Binding Protein 2|FES1|Hsp70-Interacting Protein 2|Hsp70 Interacting Protein|HSPA (Heat Shock 70kDa) Binding Protein|Cytoplasmic Cochaperone 1
- Amino acid number:84 to 359
- Storage buffer:PBS (0,2% gelatin, 0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Recombinant protein corresponding to amino acids 84-359 of human HSPBP1.
- Size:200 μg
- Pk:200 µg
Rabbit polyclonal antibody raised against partial recombinant HSPBP1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: HSPBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human