Order Entry
Denmark
ContactUsLinkComponent
Anti-SAPCD2 Rabbit Polyclonal Antibody
Anti-SAPCD2 Rabbit Polyclonal Antibody
  ABNOPAB24303
 :  Abnova
Anti-SAPCD2 Rabbit Polyclonal Antibody
  ABNOPAB24303
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    suppressor APC domain containing 2
  • Antigen symbol:
    SAPCD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    89958
  • Antigen synonyms:
    p42.3|C9orf140
  • Storage buffer:
    PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    ELEQEKEVLLQGLEMMARGRDWYQQQLQRVQERQRRLGQSRASADFGAAGSPRPLGRLLPKVQEVARC
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human SAPCD2.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against recombinant SAPCD2.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2500; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: SAPCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human