Order Entry
Denmark
ContactUsLinkComponent
Anti-PLSCR1 Mouse Monoclonal Antibody [clone: 13A6 [TEC-23]]
Anti-PLSCR1 Mouse Monoclonal Antibody [clone: 13A6 [TEC-23]]
Catalog # ABNOMAB6482
Supplier:  Abnova
Anti-PLSCR1 Mouse Monoclonal Antibody [clone: 13A6 [TEC-23]]
Catalog # ABNOMAB6482
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phospholipid Scramblase 1
  • Antigen symbol:
    PLSCR1
  • Clonality:
    Monoclonal
  • Clone:
    13A6 [TEC-23]
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Rat
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    117540
  • Storage buffer:
    15 mM sodium azide
  • Storage temperature:
    Store at 4 °C. Do not freeze. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    BALB/c mice were immunized with pooled lipid rafts isolated from RBL-2H3 cells. Hybridoma cells were obtained after fusion of SP02 mouse myeloma cells with spleen cells of immunized mice using standard procedures.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against Plscr1.

Type: Primary
Antigen: PLSCR1
Clonality: Monoclonal
Clone: 13A6 [TEC-23]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat