Order Entry
Denmark
ContactUsLinkComponent
Anti-CASP1 Rabbit Polyclonal Antibody
Anti-CASP1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18845
Supplier:  Abnova
Anti-CASP1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18845
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 1
  • Antigen symbol:
    CASP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    834
  • Antigen synonyms:
    interleukin 1-B converting enzyme|ICE|IL1B-convertase|P45|apoptosis-related cysteine peptidase interleukin 1|IL1BC|beta|IL-1 beta-converting enzyme|caspase 1|interleukin 1|convertase|caspase-1|CASP1 nirs variant 1
  • Storage buffer:
    buffer containing 0,02% sodium azide, 2% BSA
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids at N-terminus of human CASP1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of CASP1.

Recommended Dilutions: ELISA: 1:40000-1:80000; Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human