Anti-IL1B Rabbit Polyclonal Antibody
Catalog #
Product Details & Documents
Rabbit polyclonal antibody raised against a recombinant Il1b.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; ELISA: 1-2 ?g/ml
Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Documents
Literature reference - PubMed ID 17056568
Additional Product Details
TDS_ABNOPAB9535.pdf



