Anti-SCGB1D2 Goat Polyclonal Antibody
Catalog # ABNOPAB6812
Leverandør: Abnova
Specifikationer
- Antibody type:Primary
- Antigen name:Secretoglobin Family 1D, Member 2
- Antigen symbol:SCGB1D2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Cross adsorption:No
- Format:Liquid
- Gene ID:10647
- Antigen synonyms:Lipophilin B|LIPB|LPHB|LPNB
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:CTDQMSLQKRSLIAE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human SCGB1D2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifikationer
Om denne vare
Goat polyclonal antibody raised against synthetic peptide of SCGB1D2.
Recommended Dilutions: ELISA: 1:32000
Type: Primary
Antigen: SCGB1D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: