Anti-PRKAA1 Rabbit Polyclonal Antibody
Katalog # ABNOPAB19817
Leverandør: Abnova
Specifikationer
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:5562
- Antigen synonyms:AMPK|AMPKa1
- Storage buffer:1% BSA, 0,02% sodium azide
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids near internal region of human PRKAA1.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifikationer
Om denne vare
Rabbit polyclonal antibody raised against synthetic peptide of PRKAA1.
Recommended Dilutions: ELISA: 1:40000-1:80000; Western Blot: 1:200-500; Immunohistochemistry: 1:50-200
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat