Anti-PNPLA2 Goat Polyclonal Antibody
Catalog # ABNOPAB7589
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Patatin-like Phospholipase Domain Containing 2
- Antigen symbol:PNPLA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:57104
- Antigen synonyms:iPLA2zeta|TTS-2.2|ATGL|TTS2|FP17548|PEDF-R|Desnutrin
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-PARPHGPEDKDQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human PNPLA2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of PNPLA2.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.2-0.6 ?g/ml
Type: Primary
Antigen: PNPLA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human