Anti-RPGRIP1L Goat Polyclonal Antibody
ABNOPAB7600
: Abnova
- Antibody type:Primary
- Antigen name:RPGRIP1-like
- Antigen symbol:RPGRIP1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:23322
- Antigen synonyms:RPGRIP1-like |DKFZp686C0668 |MKS5|CORS3 |KIAA1005 |JBTS7
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DKENNKAKRDILK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human RPGRIP1L.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of RPGRIP1L.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: RPGRIP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human