Order Entry
Denmark
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2D1]
Anti-CD45 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2D1]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD45 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2D1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-9459-T025
  • 35-9459-T100
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase receptor type C
  • Protein tyrosine phosphatase receptor type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 2D1
  • 2D1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Frequently Bought Together