Order Entry
Denmark
ContactUsLinkComponent
Anti-CD252 (OX40 Ligand) Rat Monoclonal Antibody (APC (Allophycocyanine)) [clone: RM134L]
Anti-CD252 (OX40 Ligand) Rat Monoclonal Antibody (APC (Allophycocyanine)) [clone: RM134L]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD252 (OX40 Ligand) Rat Monoclonal Antibody (APC (Allophycocyanine)) [clone: RM134L]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-5905-U100
  • 20-5905-U025
  • Antigen symbol:
  • TNFSF4
  • TNFSF4
  • Antigen name:
  • Tumor Necrosis Factor Superfamily Member 4
  • Tumor Necrosis Factor Superfamily Member 4
  • Antigen synonyms:
  • TNFSF4|CD134L|OX-40L|TXGP1|OX40L|OX4OL|Glycoprotein Gp34|CD252|Tumor necrosis factor ligand superfamily member 4|GP34
  • TNFSF4|CD134L|OX-40L|TXGP1|OX40L|OX4OL|Glycoprotein Gp34|CD252|Tumor necrosis factor ligand superfamily member 4|GP34
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • RM134L
  • RM134L
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 22164
  • 22164
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together