Mouse IgG1 Isotype Control [clone: MOPC-21] (redFluor® 710)
Catalog # 80-4714-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:MOPC-21
- Conjugation:redFluor® 710
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8 °C
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: MOPC-21
Clonality:
Clone: MOPC-21
Conjugation: redFluor™ 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: N/A