Anti-SNUPN Rabbit Polyclonal Antibody
ABGEAP16617C
: Abgent
- Antibody type:Primary
- Antigen name:snurportin 1
- Antigen symbol:SNUPN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:10073
- Antigen synonyms:RNUT1|KPNBL|Snurportin1
- Amino acid number:200 to 228
- Storage buffer:pH 7,4
- Molecular weight:41.143 kDa
- Sequence:EEGLGEKTKLNPFKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.9
- Shipping temperature:4 °C
- Immunogen:This SNUPN antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-228 amino acids from the Central region of human SNUPN.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: SNUPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human