Anti-BLT1 Rabbit Polyclonal Antibody
Catalog # CAYM100019-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:BLT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Bovine
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Serum
- Form:lyophilized
- Amino acid number:331 to 352
- Sequence:ALEPGPSESLTASSPLKLNELN
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human BLT1 receptor amino acids 331-352 (ALEPGPSESLTASSPLKLNELN)
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: BLT1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human