Anti-Siglec 7 Mouse Monoclonal Antibody (APC (Allophycocyanine))
Do not freeze!
Type: Primary
Antigen: Siglec 7
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanine)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human,
- Catalog No:
- USBI167694
- USBI167694-100
- Antigen symbol:
- Siglec 7
- Siglec 7
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to aa19-357 from human Siglec-7, NS0-derived (Accession Q9Y286).
- Recombinant protein corresponding to aa19-357 from human Siglec-7, NS0-derived (Accession Q9Y286).
- Amino acid number:
- 19 to 357
- 19 to 357
- Form:
- liquid
- liquid
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,5%BSA, 0,09% sodium azide
- 0,5%BSA, 0,09% sodium azide
- Storage temperature:
- +4 °C
- +4 °C
Frequently Bought Together