Order Entry
Denmark
ContactUsLinkComponent
Anti-Urodilatin Rabbit Polyclonal Antibody
Anti-Urodilatin Rabbit Polyclonal Antibody
Catalog # USBIU1750-01C-100
Supplier:  US Biological
Anti-Urodilatin Rabbit Polyclonal Antibody
Catalog # USBIU1750-01C-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Urodilatin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Cross adsorption:
    No
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic rat Urodilatin
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Radio Immunoassay: 1:3000

Type: Primary
Antigen: Urodilatin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat,