Anti-Gastrin Rabbit Polyclonal Antibody
Catalog # USBIG2020-01H-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Gastrin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2
- Sequence:QGPWLEEEEEAYGWMDF
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Human Gastrin, bTG conjugated (QGPWLEEEEEAYGWMDF)
- Tested applications:Radio Immunoassay
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Radio Immunoassay: 1:5000
Type: Primary
Antigen: Gastrin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,