Anti-IgG F(c) Goat Polyclonal Antibody
AGRIAS10792
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgG F(c)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Seq:01
- Cross adsorption:No
- Storage temperature:2...8 °C
- Shipping temperature:Ambient
- Immunogen:purified human IgG
- Purification:affinity-purified
- Size:2 mg
- Pk:2 mg
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human