Order Entry
Denmark
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # AGRIAS101520
Supplier:  Agrisera
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Catalog # AGRIAS101520
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Llama
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Purified rat IgG (H&L)
  • Purification:
    affinity-purified
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Llama