Order Entry
Denmark
ContactUsLinkComponent
Anti-Steptavidin Antibody (Peroxidase)
Anti-Steptavidin Antibody (Peroxidase)
Catalog # BSBTBA1088-1
Supplier:  Boster Bio
Anti-Steptavidin Antibody (Peroxidase)
Catalog # BSBTBA1088-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Steptavidin
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Cross adsorption:
    No
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Primary
Antigen: Steptavidin
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity: