Order Entry
Denmark
ContactUsLinkComponent
General purpose gloves, ActivArmr® Medium Duty HI-VIZ™ 97-013
General purpose gloves, ActivArmr® Medium Duty HI-VIZ™ 97-013
Supplier:  Ansell
Certificates
undefined
General purpose gloves, ActivArmr® Medium Duty HI-VIZ™ 97-013
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE97-013/11
  • ANSE97-013/8
  • ANSE97-013/7
  • ANSE97-013/10
  • ANSE97-013/9
  • Model:
  • Medium duty 97-013
  • Medium duty 97-013
  • Medium duty 97-013
  • Medium duty 97-013
  • Medium duty 97-013
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyester/elastane
  • Polyester/elastane
  • Polyester/elastane
  • Polyester/elastane
  • Polyester/elastane
  • Finish:
  • Palm coated
  • Palm coated
  • Palm coated
  • Palm coated
  • Palm coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)
  • EN 388:2016 (4X32B)