Order Entry
Denmark
ContactUsLinkComponent
Syringe rack
Syringe rack
Catalog # HAMI204880
Supplier:  HAMILTON BONADUZ
undefined
Syringe rack
Catalog # HAMI204880
Supplier:  HAMILTON BONADUZ
Specifications
  • For:
    700/1700/7000 series syringes
  • Description:
    Syringe rack, holds up to 5 syringes
  • Pk:
    1
Specifications
About this item
This brushed stainless steel rack is used for clean and safe storage of syringes with plastic holder. A lip on the plastic holder protects the syringe against accidental touching and falling down. The distance of the plastic holder prevents the syringe from getting in contact with the metal plate.
  • A trough underneath prevents the soiling of worplace with liquid
  • Usable as free standing or wall model