Protective hand creams, Ligana® B Oleo-Tec
Catalog # FAWE12984001
Supplier: FAWECO HAUTSCHUTZ
Specifications
- Description:Protective hand cream, LIGANA® AQUA-tec/Creame B
- Seq:02
- Pk:100 mL
Specifications
About this item
O/W cream protects from oil-soluble substances such as oils, organic solvents, non-water soluble cooling lubricants, cutting oils, and other oily substances.
- Reduces skin irritations
- Contains glycerine and beeswax
- Silicone-free, perfumed
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904