Ordreindtastning
Denmark
ContactUsLinkComponent
VWR Chemicals Mucap test®, Salmonella identification
VWR Chemicals Mucap test®, Salmonella identification
Catalog # BLIF191500
Supplier:  BIOLIFE ITALIANA
CAS Number:  
undefined
VWR Chemicals Mucap test®, Salmonella identification
Catalog # BLIF191500
Supplier:  BIOLIFE ITALIANA
CAS Number:  
Specifications
  • Pakn:
    1 Vial
  • Beskrivelse:
    Mucap test®, 8 ml for 160 tests
Specifications
About this item
The Mucap test® allows the presumptive detection of Salmonella spp. Wood lamp with emission at 366 nm is required. Positive colonies on the plate show blue fluorescence.
  • Easy
  • Rapid