Accessories for BOD measurement lovibond BD 600, Tintometer
Supplier: Lovibond
Certificates
Certificates
About this item
Frequently Bought Together
BUFFER SOLUTION PH 6.881 @ 20C NIST 1 * 500 mL
Supplier: REAGECON
VWR®, Glass Fibre Filters
Supplier: VWR Collection
Sodium hydroxide
Supplier: WTW
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Whatman antibiotic assay discs
Supplier: Whatman products (Cytiva)
Potassium nitrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Connection Cables for IDS Sensors
Supplier: WTW
Zirconyl chloride octahydrate analytical reagent, Supelco®
Supplier: Merck
Watch glasses
Supplier: KARTELL
Wipes for general applications, Lavette Super
Supplier: PGI NONWOVEN
Gas washing bottles, DURAN®
Supplier: DWK Life Sciences
Masterflex® Direct-Reading Variable-Area Flowmeters for Air, Acrylic, Avantor®
Supplier: Avantor Fluid Handling
Potassium hydroxide 45% in aqueous solution
Supplier: Lovibond
Accessories for BOD measurement lovibond BD 600, Tintometer
Supplier: Lovibond
Dishes, evaporating
Supplier: MORGAN ADVANCED MATERIALS HALD
Sulfite lr tablets in pack of 100 1 * 100 Tablet
Supplier: Lovibond
Turbidity calibration standards, formazin, Supelco®
Supplier: Merck