Anti-HA tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 16B12]
Catalog # COBSD9-1722
Supplier: Columbia Biosciences
Specifications
- Antibody type:Tag
- Antigen name:HA tag
- Antigen symbol:HA
- Clonality:Monoclonal
- Clone:16B12
- Conjugation:DyLight® 550
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: