Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (DyLight® 488) [clone: M2]
Catalog # COBSD8-1718
Supplier: Columbia Biosciences
Specifications
- Antibody type:Tag
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Monoclonal
- Clone:M2
- Conjugation:DyLight® 488
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: