Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # COBSD5-113
Supplier: Columbia Biosciences
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Cross adsorption:Bovine, Rabbit
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat