Anti-6X His tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD1.1.10]
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
- Catalog No:
- COBSD3-1711-1MG
- COBSD3-1711-100
- Antigen symbol:
- EGLN2
- EGLN2
- Antigen name:
- egl-9 family hypoxia-inducible factor 2
- egl-9 family hypoxia-inducible factor 2
- Antigen synonyms:
- HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
- HIFPH1|HIF-PH1|EIT6|HPH-1|HPH-3|PHD1
- Antibody type:
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- AD1.1.10
- AD1.1.10
- Host:
- Mouse
- Mouse
- Gene ID:
- 112398
- 112398
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together