Anti-c-Myc tag Mouse Monoclonal Antibody (Europium 1024) [clone: 9E10]
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: Europium 1024
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
- Catalog No:
- COBSD17-1714-50
- COBSD17-1714-20
- COBSD17-1714-1MG
- Antigen symbol:
- MYC
- MYC
- MYC
- Antigen name:
- v-myc avian myelocytomatosis viral oncogene homolog
- v-myc avian myelocytomatosis viral oncogene homolog
- v-myc avian myelocytomatosis viral oncogene homolog
- Antigen synonyms:
- MRTL|bHLHe39|c-Myc|MYCC
- MRTL|bHLHe39|c-Myc|MYCC
- MRTL|bHLHe39|c-Myc|MYCC
- Antibody type:
- Tag
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Europium 1024
- Europium 1024
- Europium 1024
- Clone:
- 9E10
- 9E10
- 9E10
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4609
- 4609
- 4609
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Cross adsorption:
- none
- none
- none
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
Frequently Bought Together