Anti-WAS Rabbit Polyclonal Antibody
Katalog # USBIW0800-05
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Wiskott-Aldrich syndrome
- Antigen symbol:WAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Form:liquid
- Modification:phospho-Thr305
- Storage buffer:PBS, 50% glycerol, 1mg/ml BSA, and 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: WAS (Wiskott-Aldrich syndrome)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat