Anti-VAV1 Rabbit Polyclonal Antibody
Katalog # USBIV2121-02C
Leverandør: US Biological
Specifikationer
- Antistof type:Primary
- Antigen name:Vav 1 guanine nucleotide exchange factor
- Antigen symbol:VAV1
- Clonality:Polyclonal
- Konjugation:Unconjugated
- Vært:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Tyr174
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
- Opbevaringstemperatur:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Størrelse:50 µg
- Pakn:50 µG
Specifikationer
Om denne vare
Recommended dilution: Western Blot:1:500-1:1000
Type: Primary
Antigen: VAV1 (vav 1 guanine nucleotide exchange factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat