Anti-Trenbolone Sheep Polyclonal Antibody
Catalog # USBIT8289
Supplier: US Biological
Specifications
- Antistof type:Primary
- Antigen symbol:Trenbolone
- Clonality:Polyclonal
- Konjugation:Unconjugated
- ELISA:Yes
- Vært:Sheep
- Form:liquid
- Storage buffer:20mM PBS, 0,09% sodium azide, pH 7,2.
- Opbevaringstemperatur:Cool
- Koncentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Størrelse:100 µl
- Pakn:100 µl
Specifications
About this item
Recommended dilution: ELISA:10 ?g/ml
Type: Primary
Antigen: Trenbolone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: