Anti-Stat 6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBIS7972-40M-APC
- Antibody type:Primary
- Antigen symbol:Stat 6
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Modification:phospho-Tyr641
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: Stat 6
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human