Anti-Sprouty-related EVH1 domain-containing Protein 1, IN2 Rabbit Polyclonal Antibody
Catalog # USBIS5620-35B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Sprouty-related EVH1 domain-containing Protein 1, IN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: Sprouty-related EVH1 domain-containing Protein 1, IN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat