Anti-SK4 Goat Polyclonal Antibody
Catalog # USBIS1014-09Z
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:SK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Form:liquid
- Amino acid number:227 to 239
- Storage buffer:Tris saline buffer, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-ERQAVNATGHLSD
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 3. 75-7. 5 ?g/ml
Type: Primary
Antigen: SK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human