Anti-Oxycodone Rabbit Polyclonal Antibody
Catalog # USBIO8242-02B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Oxycodone
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Form:liquid
- Storage buffer:PBS, pH7.15, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Oxycodone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: