Anti-Nmdar1, NR1 C2 Rabbit Polyclonal Antibody
USBIN2904-36
- Antibody type:Primary
- Antigen name:NMDA Receptor NR1, Splice Variant C2
- Antigen symbol:NMDAR1, NR1 C2'
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:25 µg
- Pk:25 µG
Recommended dilution: Immunochemistry: 1:1000-1:2000; Western Blot: 1:1000
Type: Primary
Antigen: Nmdar1, NR1 C2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat