Anti-NMDA 1 Receptor, C2’ Splice Variant Rabbit Polyclonal Antibody
Catalog # USBIN2904-11
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NMDA 1 Receptor, C2’ Splice Variant
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:5mMammonium bicarbonate.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:25 µg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: NMDA 1 Receptor, C2’ Splice Variant
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat